Learn More
anti-BID Matched Antibody Pair, Ab-1, Abnova
Descrizione
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-BID (100μg); Mouse monoclonal anti-BID, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes a death agonist that heterodimerizes with either agonist BAX or antagonist BCL2. The encoded protein is a member of the BCL-2 family of cell death regulators. It is a mediator of mitochondrial damage induced by caspase-8 (CASP8); CASP8 cleaves this encoded protein, and the COOH-terminal part translocates to mitochondria where it triggers cytochrome c release. Multiple alternatively spliced transcript variants have been found, but the full-length nature of some variants has not been defined.
ELISA
Specifica
Specifica
| Antigene | BID |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BID. |
| Gene | BID |
| Alias gene | FP497/MGC15319/MGC42355 |
| Simboli geni | BID |
| Specie ospite | Mouse, Rabbit |
| Metodo di purificazione | Affinity Purified |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.