Learn More
anti-C10ORF46, Human, IP-WB Antibody Pair, Abnova™
Descrizione
Includes
Antibody pair for IP: rabbit polyclonal anti-C10orf46 (300μL), Antibody pair for WB: mouse polyclonal anti-C10orf46 (50μL)
Specifica
Specifica
| Antigene | C10orf46 |
| Applicazioni | Immunoprecipitation, Western Blot |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | C10orf46 |
| Alias gene | FLJ40409/MGC33215 |
| Simboli geni | C10orf46 |
| Specie ospite | Mouse, Rabbit |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only