missing translation for 'onlineSavingsMsg'
Learn More

anti-CBS Matched Antibody Pair, Abnova

Codice prodotto. 16151834
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16151834 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16151834 Fournisseur Abnova Code fournisseur H00000875AP11.1Set

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-CBS (100μg); Mouse monoclonal anti-CBS, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

The protein encoded by this gene acts as a homotetramer to catalyze the conversion of homocysteine to cystathionine, the first step in the transsulfuration pathway. The encoded protein is allosterically activated by adenosyl-methionine and uses pyridoxal phosphate as a cofactor. Defects in this gene can cause cystathionine β-synthase deficiency (CBSD), which can lead to homocystinuria.

ELISA

Specifications

Antigene CBS
Applicazioni ELISA
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CBS.
Gene CBS
Alias gene HIP4
Simboli geni CBS
Specie ospite Mouse, Rabbit
Metodo di purificazione Affinity Purified
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 875
Include Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CBS (100 μg) 2. Detection antibody: mouse monoclonal anti-CBS, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.