Learn More
anti-CCNE1 Matched Antibody Pair, Abnova
Descrizione
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-CCNE1 (100μg); Mouse polyclonal anti-CCNE1 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK2, whose activity is required for cell cycle G1/S transition. This protein accumulates at the G1-S phase boundary and is degraded as cells progress through S phase. Overexpression of this gene has been observed in many tumors, which results in chromosome instability, and thus may contribute to tumorigenesis. This protein was found to associate with, and be involved in, the phosphorylation of NPAT protein (nuclear protein mapped to the ATM locus), which participates in cell-cycle regulated histone gene expression and plays a critical role in promoting cell-cycle progression in the absence of pRB. Two alternatively spliced transcript variants of this gene, which encode distinct isoforms, have been described. Two additional splice variants were reported but detailed nucleotide sequence information is not yet available.
ELISA
Specifica
Specifica
| Antigene | CCNE1 |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CCNE1. |
| Gene | CCNE1 |
| Alias gene | CCNE |
| Simboli geni | CCNE1 |
| Specie ospite | Mouse, Rabbit |
| Metodo di purificazione | Affinity Purified |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.