missing translation for 'onlineSavingsMsg'
Learn More

anti-CD160, Human, IP-WB Antibody Pair, Abnova™

Codice prodotto. 16193214
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
16193214 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16193214 Fornitore Abnova N. del fornitore H00011126PW2.1Set
 Articolo soggetto a restrizioni
Spedizione stimata: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Antibody pair for IP: rabbit polyclonal anti-CD160 (300μL), Antibody pair for WB: mouse purified polyclonal anti-CD160 (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Specifications

Antigene CD160
Applicazioni Immunoprecipitation, Western Blot
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene CD160
Alias gene BY55/FLJ46513/NK1/NK28
Simboli geni CD160
Specie ospite Mouse, Rabbit
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 11126
Include Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-CD160 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-CD160 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Forma Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.