missing translation for 'onlineSavingsMsg'
Learn More

anti-FBXO6 Matched Antibody Pair, Abnova

Codice prodotto. 16163304
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16163304 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16163304 Fornitore Abnova N. del fornitore H00026270AP11.1Set
 Articolo soggetto a restrizioni
Spedizione stimata: 25-08-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-FBXO6 (100μg); Mouse monoclonal anti-FBXO6, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42kDa) which may be involved in the control of the cell cycle.

ELISA

Specifica

Antigene FBXO6
Applicazioni ELISA
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FBXO6.
Gene FBXO6
Alias gene FBG2/FBS2/FBX6/Fbx6b
Simboli geni FBXO6
Specie ospite Mouse, Rabbit
Metodo di purificazione Affinity Purified
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 26270
Include Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FBXO6 (100 μg) 2. Detection antibody: mouse monoclonal anti-FBXO6, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.