Learn More
anti-FCER2 Ab-1 Matched Antibody Pair, Abnova™
Descrizione
Includes
Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FCER2 (100μg); Detection antibody: mouse purified polyclonal anti-FCER2 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Host Species: Rabbit, Mouse
Cross Reactivity: Human
ELISA Pair
Specifica
Specifica
| Antigene | FCER2 |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FCER2. |
| Gene | FCER2 |
| Alias gene | CD23/CD23A/CLEC4J/FCE2/IGEBF |
| Simboli geni | FCER2 |
| Specie ospite | Mouse, Rabbit |
| Metodo di purificazione | Affinity Purified |
| Vedi altri risultati |