Learn More
anti-FGA, F2 Protein Protein Interaction Antibody Pair, Abnova™
Descrizione
Includes
FGA rabbit purified polyclonal antibody (20μg); F2 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the FGA protein, and the other against the F2 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifica
Specifica
| Antigene | FGA |
| Applicazioni | In situ PLA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Gene | FGA |
| Alias gene | Fib2/MGC119422/MGC119423/MGC119425 |
| Simboli geni | FGA |
| Specie ospite | Mouse, Rabbit |
| Quantità | 1 Set |
| Status giuridico | RUO |
| Vedi altri risultati |