Learn More
anti-FLG1 Matched Antibody Pair, Abnova
Descrizione
Includes
Rabbit affinity purified polyclonal anti-FGL1 (100μg); Mouse monoclonal anti-FGL1, IgG2bκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen β- and γ- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.
ELISA
Specifica
Specifica
| Antigene | FGL1 |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FGL1. |
| Gene | FGL1 |
| Alias gene | HFREP1/HP-041/LFIRE1/MGC12455 |
| Simboli geni | FGL1 |
| Specie ospite | Mouse, Rabbit |
| Metodo di purificazione | Affinity Purified |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.