Learn More
anti-GNL3, Human, Matched Antibody Pair, Abnova™
Descrizione
Includes
Capture antibody: mouse monoclonal anti-GNL3, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-GNL3, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifica
Specifica
| Antigene | GNL3 |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Biotin |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GNL3. |
| Gene | GNL3 |
| Alias gene | C77032/E2IG3/MGC800/NS |
| Simboli geni | GNL3 |
| Specie ospite | Mouse |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only