Learn More
IGKC, Human, IP-WB Antibody Pair (PW1), Abnova™
Descrizione
Includes
Antibody pair for IP: mouse monoclonal anti-IGKC (300μg), Antibody pair for WB: rabbit polyclonal anti-IGKC (50μL)
Specifica
Specifica
| Antigene | IGKC |
| Applicazioni | Immunoprecipitation, Western Blot |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | IGKC |
| Alias gene | HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 |
| Simboli geni | IGKC |
| Specie ospite | Mouse, Rabbit |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only