Learn More
anti-IGKC, Human, Matched Antibody Pair, Abnova™
Descrizione
Includes
Capture antibody: rabbit MaxPab affinity purified polyclonal anti-IGKC (100μg) , Detection antibody: mouse purified polyclonal anti-IGKC (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifica
Specifica
| Antigene | IGKC |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IGKC. |
| Gene | IGKC |
| Alias gene | HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 |
| Simboli geni | IGKC |
| Specie ospite | Mouse, Rabbit |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only