Learn More
IL-4 Rat anti-Mouse, PE-CF594, Clone: 11B11, BD
Descrizione
The 11B11 monoclonal antibody specifically binds to mouse Interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 prepared from the supernatant of Phorbol 12-Myristate 13-Acetate (PMA)-stimulated EL-4 cells. The 11B11 antibody is reportedly a neutralizing antibody.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Specifica
Specifica
| Antigene | IL-4 |
| Applicazioni | Flow Cytometry |
| Classificazione | Monoclonal |
| Clone | 11B11 |
| Concentrazione | 0.2mg/mL |
| Coniugato | PE-CF594 |
| Descrizione | IL4: Interleukin-4; BSF-1; B-cell growth factor 1; BCGF-1 |
| Formulazione | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Specie ospite | Rat |
| Immunogeno | Partially Purified Mouse IL-4 |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.