Learn More
IL13, Human, Matched Antibody Pair (AP44), Abnova™
Descrizione
Includes
Capture antibody: mouse monoclonal anti-IL13 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-IL13, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifica
Specifica
| Antigene | IL13 |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Biotin |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL13. |
| Gene | IL13 |
| Alias gene | ALRH/BHR1/IL-13/MGC116786/MGC116788/MGC116789/P600 |
| Simboli geni | IL13 |
| Specie ospite | Mouse |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only