Learn More
anti-KHK Matched Antibody Pair, Abnova
Codice prodotto. 16036428
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16036428 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16036428 Fornitore Abnova N. del fornitore H00003795AP11.1Set
Please to purchase this item. Need a web account? Register with us today!
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-KHK (100μg); Mouse monoclonal anti-KHK, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified.
ELISA
Specifica
Antigene KHK
Applicazioni ELISA
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KHK.
Gene KHK
Simboli geni KHK
Specie ospite Mouse, Rabbit
Metodo di purificazione Affinity Purified
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 3795
Include Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-KHK (100 μg) 2. Detection antibody: mouse monoclonal anti-KHK, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG2a