missing translation for 'onlineSavingsMsg'
Learn More

CHRM1 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marca:  Invitrogen PA518570

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 11386543

  • 473.00€ / 100µg
Spedizione stimata: 09-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This antibody is predicted to react with canine, mouse and rat based on sequence homology.

CHRM1 Polyclonal antibody specifically detects CHRM1 in Mouse samples. It is validated for Western Blot.
Specifica

Specifica

CHRM1
Polyclonal
Unconjugated
CHRM1
acetylcholine receptor; acetylcholine receptor, muscarinic 1; AChR M1; cholinergic receptor; cholinergic receptor muscarinic 1; Cholinergic receptor, muscarin 1; cholinergic receptor, muscarinic 1; cholinergic receptor, muscarinic 1, CNS; CHRM1; Chrm-1; HM1; M1; M1 mAChR; m1 muscarinic acetylcholine receptor protein; M1R; MGC30125; muscarinic acetylcholine receptor M1; muscarinic receptor M1
Goat
Antigen affinity chromatography
RUO
12669
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P12657
CHRM1
Synthetic peptide sequence (DKRRWRKIPKRPGSVH) corresponding to the C-terminus amino acids of CHRM1
100 μg
Primary
Mouse
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali