missing translation for 'onlineSavingsMsg'
Learn More

MCM2, Human, IP-WB Antibody Pair (PW1), Abnova™

Codice prodotto. 16096768
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
16096768 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16096768 Fornitore Abnova N. del fornitore H00004171PW1.1Set
 Articolo soggetto a restrizioni
Spedizione stimata: 26-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes

Antibody pair for IP: rabbit polyclonal anti-MCM2 (300μL), Antibody pair for WB: rabbit polyclonal anti-MCM2 (50μL)

For immunoprecipitation and to detect the precipitated protein in western blot.

Spezifikation

Antigene MCM2
Applicazioni Immunoprecipitation, Western Blot
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene MCM2
Alias gene BM28/CCNL1/CDCL1/D3S3194/KIAA0030/MGC10606/MITOTIN/cdc19
Simboli geni MCM2
Specie ospite Rabbit
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 4171
Include Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-MCM2 (300 ul) 2. Antibody pair for WB: rabbit polyclonal anti-MCM2 (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Forma Liquid
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.