missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor 680, Invitrogen™
Descrizione
Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor 680 conjugate, A31562, from Invitrogen™. Species Reactivity: Mouse; Applications: Immunocytochemistry, Immunofluorescence, Western Blot.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Host Species: Goat
Cross Reactivity: Mouse
Isotype: IgG
Immunogen: IgG gamma 1
Formulation: 250μL purified antibody (2mg/mL) in PBS and 5mM sodium azide; pH 7.5
This secondary antibody is designed for fluorescent Western blot detection on various near-infrared fluorescence instruments. This antibody can be used for multi-color and multiplexing detection when using other antibodies conjugated to compatible Alexa Fluor dyes and wavelengths. Other applications of this antibody include immunofluorescent and fluorescent imaging applications when using instrumentation with appropriate excitation and detection capabilities.
Host Species: Goat
Cross Reactivity: Mouse
Isotype: IgG
Immunogen: IgG gamma 1
Formulation: 250μL purified antibody (2mg/mL) in PBS and 5mM sodium azide; pH 7.5
This secondary antibody is designed for fluorescent Western blot detection on various near-infrared fluorescence instruments. This antibody can be used for multi-color and multiplexing detection when using other antibodies conjugated to compatible Alexa Fluor dyes and wavelengths. Other applications of this antibody include immunofluorescent and fluorescent imaging applications when using instrumentation with appropriate excitation and detection capabilities.
Specifica
Specifica
| Antigene | Mouse IgG1 (Alexa Fluor™ 680) |
| Classificazione | Polyclonal |
| Coniugato | Alexa Fluor 680 |
| Specie ospite | Goat |
| Immunogeno | IgG gamma 1 |
| Primario o secondario | Secondary |
| Target Species | Mouse |
| Content And Storage | Store at 4°C, store in dark |
| Forma | Purified |
| Isotype | IgG |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?