Learn More
Mycoplasma genitalium Monoclonal Antibody (A46J), Invitrogen™
Mouse Monoclonal Antibody
Marca: Thermo Scientific MA518227
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody is specific to M. genitalium and does not cross-react with Mycoplasma hominis, Mycoplasma pneumoniae, Ureaplasma urealyticum, Herpes virus type 2, Chlamydia trachomatis, Neisseria gonorrhoeae, or human cells. Purity is >90%. Recommended pairs for ELISA: MA5-18226 or MA5-18228 (capture) and MA5-18227 (conjugated, detection). MA5-18227 can also be used as a capture antibody.
Mycoplasma genitalium Monoclonal antibody specifically detects Mycoplasma genitalium in Bacteria samples. It is validated for ELISA, Immunocytochemistry.Specifica
Mycoplasma genitalium | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry | |
A46J | |
Unconjugated | |
M. genitalium; mycoplasmen | |
Mycoplasma genitalium native protein | |
100 μg | |
Primary | |
4° C | |
Liquid |