Learn More
PPP2R3B, Human, IP-WB Antibody Pair (PW2), Abnova™
Codice prodotto. 16113344
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16113344 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16113344 Fornitore Abnova N. del fornitore H00028227PW2.1Set
Please to purchase this item. Need a web account? Register with us today!
Includes
Antibody pair for IP: mouse monoclonal anti-PPP2R3B (300μg), Antibody pair for WB: rabbit polyclonal anti-PPP2R3B (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifica
Antigene PPP2R3B
Applicazioni Immunoprecipitation, Western Blot
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene PPP2R3B
Alias gene NY-REN-8/PPP2R3L/PPP2R3LY/PR48
Simboli geni PPP2R3B
Specie ospite Mouse, Rabbit
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 28227
Include Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-PPP2R3B (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-PPP2R3B (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Forma Liquid
For Research Use Only