missing translation for 'onlineSavingsMsg'
Learn More

anti-RAD51L3 Matched Antibody Pair, Abnova

Codice prodotto. 16112544
Cambia vista
Click to view available options
Quantità:
1 Set
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16112544 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16112544 Fornitore Abnova N. del fornitore H00005892AP21.1Set

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-RAD51L3 (100μg); Mouse purified polyclonal anti-RAD51L3 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, which are known to be involved in the homologous recombination and repair of DNA. This protein forms a complex with several other members of the RAD51 family, including RAD51L1, RAD51L2, and XRCC2. The protein complex formed with this protein has been shown to catalyze homologous pairing between single- and double-stranded DNA, and is thought to play a role in the early stage of recombinational repair of DNA. Several alternatively spliced transcript variants of this gene have been described, but the biological validity of some of them has not been determined.

ELISA

Specifica

Antigene RAD51L3
Applicazioni ELISA
Classificazione Monoclonal/Polyclonal
Coniugato Unconjugated
Descrizione This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RAD51L3.
Gene RAD51L3
Alias gene HsTRAD/R51H3/RAD51D/Trad
Simboli geni RAD51L3
Specie ospite Mouse, Rabbit
Metodo di purificazione Affinity Purified
Quantità 1 Set
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 5892
Include Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RAD51L3 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-RAD51L3 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.