missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF183 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA523862
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody is predicted to react with bovine based on sequence homology.
Specifica
RNF183 | |
Polyclonal | |
RNF183 | |
RING finger protein 183 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
138065, 76072 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q96D59, Q8QZS5 | |
RNF183 | |
KLH conjugated synthetic peptide between 86-114 amino acids from the central region of human RNF183 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |