Learn More
anti-RPL14, Human, IP-WB Antibody Pair, Abnova™
Descrizione
Includes
Antibody pair for IP: rabbit polyclonal anti-RPL14 (300μL), Antibody pair for WB: mouse purified polyclonal anti-RPL14 (50μg)
Specifica
Specifica
| Antigene | RPL14 |
| Applicazioni | Immunoprecipitation, Western Blot |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | RPL14 |
| Alias gene | CAG-ISL-7/CTG-B33/L14/MGC88594/RL14/hRL14 |
| Simboli geni | RPL14 |
| Specie ospite | Mouse, Rabbit |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only