missing translation for 'onlineSavingsMsg'
Learn More

SLC27A5 Monoclonal Antibody (4B11C10), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA517175

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 15308140

  • 462.00€ / 100µL
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

MA5-17175 targets SLC27A5 in indirect ELISA, IHC and WB applications and shows reactivity with Human and Mouse samples. The MA5-17175 immunogen is purified recombinant fragment of human SLC27A5 (amino acids: 508-570 ) expressed in E. Coli. MA5-17175 detects SLC27A5 which has a predicted molecular weight of approximately 75.4kDa.

SLC27A5 Monoclonal antibody specifically detects SLC27A5 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.
Specifica

Specifica

SLC27A5
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q4LDG0, Q9Y2P5
SLC27A5
Purified recombinant fragment of human SLC27A5 (amino acids: 508-570 ) expressed in E. Coli.
100 μg
Primary
Human, Mouse
Antibody
IgG1
ELISA, Immunohistochemistry (Paraffin), Western Blot
4B11C10
Unconjugated
SLC27A5
ACSB; ACSVL6; BA-CoA ligase; BACS; BAL; bile acid CoA ligase; bile acid-CoA ligase; Bile acyl-CoA synthetase; cholate--CoA ligase; FACVL3; FATP5; FATP-5; Fatty acid transport protein 5; fatty-acid-Coenzyme A ligase, very long-chain 3; FLJ22987; rBAL-1; Slc27a5; solute carrier family 27 (fatty acid transporter), member 5; solute carrier family 27 member 5; ve; Very long-chain acyl-CoA synthetase homolog 2; very long-chain acyl-CoA synthetase-related protein; very long-chain acyl-Coenzyme A dehydrogenase synthase related; Vlacsr; VLACS-related; VLCSH2; VLCS-H2
Mouse
Protein G
RUO
10998, 26459
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali