missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Smooth Muscle Actin Rabbit anti-Human, Polyclonal, Invitrogen™

Codice prodotto. 13497097
Cambia vista
Click to view available options
Quantità:
500 μL
Dimensione della confezione:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
13497097 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 13497097 Fornitore Invitrogen N. del fornitore PA532587

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

A suggested positive control is colon or small intestine tissue.

Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Alpha-Smooth Muscle Actin
Applicazioni Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Gene ACTA2
N. accesso geni P62736
Alias gene AAT6, ACTSA, MYMY5, actin, aortic smooth muscle, alpha 2 actin, alpha-actin-2, alpha-cardiac actin, cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, alpha-sma
Simboli geni ACTA2
Specie ospite Rabbit
Immunogeno Synthetic peptide near the N-terminus of human smooth muscle actin-alpha.
Metodo di purificazione Antigen affinity chromatography
Quantità 500 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 59
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.