Learn More
anti-TYMS Matched Antibody Pair, Abnova
Descrizione
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-TYMS (100μg); Mouse purified polyclonal anti-TYMS (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Thymidylate synthase catalyzes the methylation of deoxyuridylate to deoxythymidylate using 5,10-methylenetetrahydrofolate (methylene-THF) as a cofactor. This function maintains the dTMP (thymidine-5-prime monophosphate) pool critical for DNA replication and repair. The enzyme has been of interest as a target for cancer chemotherapeutic agents. It is considered to be the primary site of action for 5-fluorouracil, 5-fluoro-2-prime-deoxyuridine, and some folate analogs. Expression of this gene and that of a naturally occurring antisense transcript rTSα (GeneID:55556) vary inversely when cell-growth progresses from late-log to plateau phase.
ELISA
Specifica
Specifica
| Antigene | TYMS |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TYMS. |
| Gene | TYMS |
| Alias gene | HsT422/MGC88736/TMS/TS/TSase |
| Simboli geni | TYMS |
| Specie ospite | Mouse, Rabbit |
| Metodo di purificazione | Affinity Purified |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.