Learn More
anti-UROS Matched Antibody Pair, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-UROS (100μg); Mouse purified polyclonal anti-UROS (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The protein encoded by this gene catalyzes the fourth step of porphyrin biosynthesis in the heme biosynthetic pathway. Defects in this gene cause congenital erythropoietic porphyria (Gunther's disease).
ELISA
Specifications
Specifications
| Antigene | UROS |
| Applicazioni | ELISA |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human UROS. |
| Gene | UROS |
| Simboli geni | UROS |
| Specie ospite | Mouse, Rabbit |
| Metodo di purificazione | Affinity Purified |
| Quantità | 1 Set |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.