missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein O Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA551853

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15719810

  • 512.00€ / 100µL
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: IKKLVYPPGF MGLAASLYYP QQAIVFAQVS GERLYDWGLR GYIVIEDLWK ENFQKPGNVK NSPGTK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 80%.

Apolipoprotein O Polyclonal antibody specifically detects Apolipoprotein O in Human samples. It is validated for Immunocytochemistry, Western Blot.
Specifica

Specifica

Apolipoprotein O
Polyclonal
Unconjugated
APOO
0610008C08Rik; 1110019O03Rik; Apolipoprotein O; APOO; brain my025; FAM121B; family with sequence similarity 121B; MGC4825; Mic23; MIC26; MICOS complex subunit Mic23; MICOS complex subunit MIC26; My025; MYO25; Protein FAM121B; RGD1565289; UNQ1866/PRO4302
Rabbit
Antigen affinity chromatography
RUO
79135
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
0.08 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BUR5
APOO
Recombinant protein corresponding to Human Apolipoprotein O
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only