missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APS Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 11582500
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11582500 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11582500 Fornitore Invitrogen™ N. del fornitore PA517608

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other adaptor/docking proteins.

APS (adapter protein with Pleckstrin homology and Src homology 2 domains) a member of the Lnk family, is an adaptor protein that is involved in B-cell signaling, insulin signaling, cytoskeletal reorganization. It is tyrosine phosphorylated by JAK2, KIT and other kinases during B-cell receptor stimulation of many different cytokines, chemokines and leukokines. APS has been shown to inhibit the JAK-STAT pathway in collaboration with c-Cbl. It is located in the cytoplasm in pre-PDGF (platelet derived growth factor) stimulated cells and localizes to the plasma membrane and peripheral regions upon PDGF stimulation. This protein is expressed in B-cells and mast cells when detected in haematopoietic cell lines yet is also expressed in many different tissues including spleen, prostrate, testis, uterus, small intestine and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifica

Antigene APS
Applicazioni Immunohistochemistry (Paraffin), Western Blot
Classificazione Polyclonal
Concentrazione 78 μg/mL
Coniugato Unconjugated
Formulazione 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene SH2B2
N. accesso geni O14492, Q9JID9
Alias gene adapter protein with pleckstrin homology and Src homology 2 domains; adaptor protein with pleckstrin homology and src homology 2 domains; Alpha-1-antitrypsin; Aps; SH2 and PH domain-containing adapter protein APS; SH2B adapter protein 2; SH2B adaptor protein 2; Sh2b2
Simboli geni SH2B2
Specie ospite Rabbit
Immunogeno Synthetic peptide corresponding to residues near the amino-terminus of human APS.
Metodo di purificazione Antigen affinity chromatography
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 10603, 23921
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.