missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AQR Polyclonal Antibody
Descrizione
This target displays homology in the following species: Horse: 79%; Human: 100%.
Involved in pre-mRNA splicing as component of the spliceosome (PubMed:11991638, PubMed:25599396, PubMed:28502770, PubMed:28076346). Intron-binding spliceosomal protein required to link pre-mRNA splicing and snoRNP (small nucleolar ribonucleoprotein) biogenesis (PubMed:16949364). Plays a key role in position-dependent assembly of intron-encoded box C/D small snoRNP, splicing being required for snoRNP assembly (PubMed:16949364). May act by helping the folding of the snoRNA sequence. Binds to intron of pre-mRNAs in a sequence-independent manner, contacting the region between snoRNA and the branchpoint of introns (40 nucleotides upstream of the branchpoint) during the late stages of splicing (PubMed:16949364). Has ATP-dependent RNA helicase activity and can unwind double-stranded RNA molecules with a 3' overhang (in vitro) (PubMed:25599396). [UniProt].
Specifica
Specifica
| Antigene | AQR |
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.5 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | AQR |
| N. accesso geni | O60306 |
| Alias gene | Aqr; aquarius; aquarius homolog; aquarius intron-binding spliceosomal factor; AW495846; fSAP164; functional spliceosome-associated protein 164; IBP160; intron-binding protein aquarius; intron-binding protein of 160 kDa; Kiaa0560; mKIAA0560; RNA helicase aquarius |
| Simboli geni | AQR |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?