missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arfolitixorin, TRC
Specifications
Specifications
| Materiale o nome chimico | (6aR)-5,10-Methylenetetrahydrofolic Acid |
| CAS | 31690-11-6 |
| Formula molecolare | C20 H23 N7 O6 |
| Quantità | 100 mg |
| Sinonimo | Arfolitixorin, L-Glutamic acid, N-[4-[(6aR)-3-amino-1,2,5,6,6a,7-hexahydro-1-oxoimidazo[1,5-f]pteridin-8(9H)-yl]benzoyl]-, Glutamic acid, N-[p-(3-amino-5,6,6a,7-tetrahydro-1-hydroxyimidazo[1,5-f]pteridin-8(9H)-yl)benzoyl]-, L-(+)- (8CI), L-Glutamic acid, N-[4-(3-amino-1,2,5,6,6a,7-hexahydro-1-oxoimidazo[1,5-f]pteridin-8(9H)-yl)benzoyl]-, (R)-, N-[4-[(6aR)-3-Amino-1,2,5,6,6a,7-hexahydro-1-oxoimidazo[1,5-f]pteridin-8(9H)-yl]benzoyl]-L-glutamic acid, (6R)-5,10-Methylene-5,6,7,8-tetrahydrofolic acid, (6R)-5,10-Methylenetetrahydrofolate, 5,10-Methylene-(6R)-tetrahydrofolic acid, Arfolitixorin, L-(+)-Methylenetetrahydrofolic acid, d-N5,N10-Methylene-L-tetrahydrofolic acid, (2S)-2-[4-[(6aR)-3-Amino-1-oxo-1,2,5,6,6a,7-hexahydroimidazo[1,5-f]pteridin-8(9H)-yl]benzamido]pentanedioic acid, (6aR)-5,10-Methylenetetrahydrofolic acid |
| InChI Formula | InChI=1S/C20H23N7O6/c21-20-24-16-15(18(31)25-20)27-9-26(8-12(27)7-22-16)11-3-1-10(2-4-11)17(30)23-13(19(32)33)5-6-14(28)29/h1-4,12-13H,5-9H2,(H,23,30)(H,28,29)(H,32,33)(H4,21,22,24,25,31)/t12-,13+/m1/s1 |
| SMILES | NC1=NC(=O)C2=C(NC[C@@H]3CN(CN23)c4ccc(cc4)C(=O)N[C@@H](CCC(=O)O)C(=O)O)N1 |
| IUPAC Name | (2S)-2-[[4-[(6aR)-3-amino-1-oxo-4,5,6,6a,7,9-hexahydroimidazo[1,5-f]pteridin-8-yl]benzoyl]amino]pentanedioic acid |
| Molecular Weight (g/mol) | 457.44 |
| Peso formulazione | 457.171 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?