Learn More
ARID1A Monoclonal Antibody (CL3595), Invitrogen™
Mouse Monoclonal Antibody
Marca: Thermo Scientific MA524658
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: PGLGNVAMGP RQHYPYGGPY DRVRTEPGIG PEGNMSTGAP QPNLMPSNPD SGMYSPSRYP PQQQQQQQQR HDSYGNQFST QGTPSGSPFP SQQTTMYQQQ QQNYK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 88%. Binds to an epitope located within the peptide sequence VAMGPRQHYPYGGPY as determined by overlapping synthetic peptides.
ARID1A Monoclonal antibody specifically detects ARID1A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
ARID1A | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14497 | |
ARID1A | |
Recombinant protein corresponding to Human ARID1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
CL3595 | |
Unconjugated | |
ARID1A | |
1110030E03Rik; ARID domain-containing protein 1A; ARID1A; AT rich interactive domain 1A (Swi1 like); AT rich interactive domain 1A (SWI-like); AT-rich interaction domain 1A; AT-rich interactive domain-containing protein 1A; B120; BAF250; Baf250a; BM029; brain protein 120; BRG1-associated factor 250; BRG1-associated factor 250a; C1orf4; chromatin remodeling factor p250; CSS2; ELD; h BM029; hELD; hOSA1; MRD14; Osa homolog 1; Osa1; OSA1 nuclear protein; P270; Smarcf1; SWI/SNF complex protein p270; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily f, member 1; SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatin subfamily F member 1; Swi1 like; SWI-like protein; SWI-SNF complex protein p270 | |
Mouse | |
Protein A | |
RUO | |
8289 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only