Learn More
Arylex, TRC
Codice prodotto. 30340916
Cambia vista
Click to view available options
Quantità:
2.5 mg
Dimensione della confezione:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30340916 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30340916 Fornitore Toronto Research Chemicals N. del fornitore TRCA7948902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Halauxifen-methyl
CAS 943831-98-9
Formula molecolare C14 H11 Cl2 F N2 O3
Quantità 2.5 mg
Sinonimo Arylex, Arylex active, Halauxifen-methyl, XDE 729
InChI Formula InChI=1S/C14H11Cl2FN2O3/c1-21-13-7(15)4-3-6(11(13)17)9-5-8(18)10(16)12(19-9)14(20)22-2/h3-5H,1-2H3,(H2,18,19)
SMILES COC(=O)c1nc(cc(N)c1Cl)c2ccc(Cl)c(OC)c2F
IUPAC Name methyl 4-amino-3-chloro-6-(4-chloro-2-fluoro-3-methoxyphenyl)pyridine-2-carboxylate
Molecular Weight (g/mol) 345.15
Peso formulazione 344.0131