Learn More
Hirschmann™ Aspirette™ Suction Unit Set (x 3)
Descrizione
- For use with aspirette™ vacuum tweezers
- Includes 3 suction nozzles and suction cups with 3.2, 6 and 10 mm diameters
- Accessory item
Specifica
Specifica
| Product Type | Suction unit set |
| Da utilizzare con (applicazione) | Picking up, positioning plane objects |
| Diametro (sistema metrico) | 3.2, 6, 10 mm |