Description
- Light and breathable safety glove
- Particularly flexible and sensitive glove for multiple areas of application
- Touchscreen capability
- Certified in line with Oeko-Tex Standard 100
- Good grip in dry and slightly damp areas
- High breathability thanks to open-pored coating that reduces perspiration inside the glove
- Good tactile feel when carrying out precision work
- Perfect fit thanks to the 'slim fit' design and elastane in the liner
Specifications
Specifications
| Certificazione/Conformità | DIN EN 16350, EN 388 (4121X) |
| Coating Material | NBR MicroFoam |
| Colore | Blue, Anthracite |
| Monouso | Reusable |
| Finitura esterna | Smooth |
| Hand | Ambidestri |
| Uso per laboratorio/camera controllata | Not tested |
| Latex Free | Sì |
| Materiale | Polyamide, Carbon Fiber, Elastane |
| Powdered | Powder Free |
| Show More |