Learn More
uvex Athletic Lite ESD Safety Glove
Codice prodotto. p-7228493 Sfoglia Tutto Uvex Prodotti
Cambia vista
Click to view available options
Size:
6
7
8
9
10
11
Dimensione della confezione:
10 paia
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Size unitSize
17707569 6 10 paia
17717569 7 10 paia
17727569 8 10 paia
17737569 9 10 paia
17747569 10 10 paia
17757569 11 10 paia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 17707569 Fornitore uvex N. del fornitore 6003506
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Particularly lightweight safety glove with ESD and touchscreen function.
  • Light and breathable safety glove
  • Particularly flexible and sensitive glove for multiple areas of application
  • Touchscreen capability
  • Certified in line with Oeko-Tex Standard 100
  • Good grip in dry and slightly damp areas
  • High breathability thanks to open-pored coating that reduces perspiration inside the glove
  • Good tactile feel when carrying out precision work
  • Perfect fit thanks to the 'slim fit' design and elastane in the liner
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità DIN EN 16350, EN 388 (4121X)
Coating Material NBR MicroFoam
Colore Blue, Anthracite
Monouso Reusable
Finitura esterna Smooth
Hand Ambidestri
Uso per laboratorio/camera controllata Not tested
Latex Free
Materiale Polyamide, Carbon Fiber, Elastane
Powdered Powder Free
Product Type ESD Gloves
Size 6
Sterilità Non-sterile