missing translation for 'onlineSavingsMsg'
Learn More

ATP5A1 Mouse anti-Human, Mouse, Non-human primate, Rat, Clone: 1B10H3, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66037-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897545

  • 325.38€ / 150µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This antibody recognizes the endogenous ATP5A1 protein in lysates from various cell lines and tissues. The predicted MW of ATP5A1 is 60 kDa, while it undergoes the transit peptide cleavage to become a mature form around 50-55 kDa. Several isoforms of ATP5

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, using an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the alpha subunit of the catalytic core. Alternatively spliced transcript variants encoding the different isoforms have been identified. Pseudogenes of this gene are located on chromosomes 9, 2, and 16.
Specifica

Specifica

ATP5A1
Monoclonal
0.95 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15999, P25705, Q03265
ATP5A1
ATP5A1 Fusion Protein Ag8119
150 μL
Primary
Rat, Non-human Primate, Mouse, Human
Antibody
IgG2b
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry
1B10H3
Unconjugated
ATP5A1
ATP5A, ATP5A1, ATP5AL2, ATPM, hATP1, MOM2, OMR, ORM
Mouse
Protein A
RUO
11946, 498, 65262
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali