missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATP6V1C1 Polyclonal antibody specifically detects ATP6V1C1 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigene | ATP6V1C1 |
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Diluizione | Western Blot 1:500-1:2000 |
| Formulazione | PBS with 50% glycerol, pH7.3. |
| Alias gene | ATP6CH+-transporting ATPase chain C, vacuolar, ATP6Dsubunit C of vacuolar proton-ATPase V1 domain, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 42kD, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C, isoform 1, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1, FLJ20057, H(+)-transporting two-sector ATPase, subunit C, vacuolar ATP synthase subunit C, vacuolar proton pump C subunit, Vacuolar proton pump subunit C 1, vacuolar proton pump, 42-kD subunit, vacuolar proton-ATPase, subunit C, VI domain, VATCH+ -ATPase C subunit, V-ATPase C subunit, V-ATPase subunit C 1, Vma5, V-type proton ATPase subunit C 1 |
| Specie ospite | Rabbit |
| Immunogeno | A synthetic peptide corresponding to a sequence within amino acids 300-382 of human ATP6V1C1 (NP_001686.1). LRVFVESVLRYGLPVNFQAMLLQPNKKTLKKLREVLHELYKHLDSSAAAIIDAPMDIPGLNLSQQEYYPYVYYKIDCNLLEFK |
| Metodo di purificazione | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?