missing translation for 'onlineSavingsMsg'
Learn More

ATP8B1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA553840

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15791771

  • 516.00€ / 100µL
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: KIWVLTGDKK ETAENIGFAC ELLTEDTTIC YGEDINSLLH ARMENQRNRG GVYAKFAPPV QESFFPPGGN RALII Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

ATP8B1 Polyclonal antibody specifically detects ATP8B1 in Human samples. It is validated for Immunohistochemistry.
Specifica

Specifica

ATP8B1
Polyclonal
Unconjugated
ATP8B1
ATP8B1; ATPase class I type 8B member 1; ATPase phospholipid transporting 8B1; ATPase, aminophospholipid transporter, class I, type 8B, member 1; ATPase, class I, type 8B, member 1; ATPIC; BRIC; E1-E2 ATPase; familial intrahepatic cholestasis type 1; FIC1; ICP1; P4-ATPase flippase complex alpha subunit ATP8B1; PFIC; PFIC1; phospholipid-transporting ATPase IC; probable phospholipid-transporting ATPase IC
Rabbit
Antigen affinity chromatography
RUO
5205
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43520
ATP8B1
Recombinant protein corresponding to Human ATP8B1
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only