Learn More
Axygen™ EasyPress Sealing Mat and Film Applicator
Product Code. 12683926 Shop All Axygen Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12683926 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12683926 Supplier Axygen™ Supplier No. ITEPR
  • ONLINE EXCLUSIVE PRICE
    2640.00€
    1575.50€ / pezzi 1
    Risparmia 1064.50€
    40% Sconto
View all promotions
Spedizione stimata: 09-12-2026
Provides simple and effective application of Axymats and ImpermaMats
Axygen™ EASY PRESS is a manual device which provides simple and effective application of Axymats and/or Impermamats to 24, 48, 96 or 384 well microplates, deep well plates, microtiter tube systems and PCR plates.The EASY PRESS can be quickly adjusted to handle any size plate of varying heights (384 PCR to 2mL deep well blocks) using the large thumbwheel.
  • Suitable for deep well plates, PCR microplates in 96 and 384 formats
  • Flexible-can be used with plates of various heights and types
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) Axymats, deep well plates, microplates, pcr plates
    No. per Pack 1 Ea.
    Product Type Sealer
    Quantity 1 Ea.