Learn More
Axygen™ Sealing Film Roller
Product Code. 11370215 Shop All Axygen Scientific Products
Change view
Click to view available options
Quantity:
1 Unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370215 1 Unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11370215 Supplier Axygen™ Supplier No. PCRSPROLLER
  • ONLINE EXCLUSIVE PRICE
    64.65€
    43.64€ / pezzi 1
    Risparmia 21.01€
    32% Sconto
Spedizione stimata: 09-02-2027
Ideal for the application of all PlateMax™ sealing films
Axygen™ sealing film roller is a durable and efficient roller ideal for applying sealing films to any plate. Sturdy plastic handle and semi-hard roller head are ideal for complete and easy applications of film.
  • Ideal for the application of all PlateMaxTM sealing films
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) PlateMax™ Sealing Films
    No. per Pack 1 Ea.
    Product Type Sealing Film Roller
    Quantity 1 Unit