Learn More
Axygen™ Wellpro™ Robotic Tips
Clear tips
Marca: Axygen TWP-200-R
Dettagli aggiuntivi : Peso : 10.90000kg
Descrizione
- Racked
- 200μL capacity
- Presterilized tips available
- Grey racks
Order Info
30++l
200++l
Specifica
200 μL | |
Clear | |
Round | |
Non-ANSI | |
Non-pyrogenic, used for stringent specifications under strict process controls | |
ProGroup Wellpro™ 3000 | |
Automation |
Non-sterile | |
Gray | |
PP (Polypropylene) | |
DNase, Rnase Free | |
ProGroup Wellpro™ 3000 | |
Rack |