missing translation for 'onlineSavingsMsg'
Learn More

AZD-6280, MedChemExpress

Product Code. p-200100897
Encompass_Preferred
Change view
Click to view available options
Quantità:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
16490408 5 mg 5mg
16457948 10 mg 10mg
16421889 1 mg 1mg
16400616 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490408 Supplier Medchem Express Supplier No. HY19872

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

MedChemExpress AZD-6280 is a selective GABAA(α2/3) receptor modulator, used for treatment of generalized anxiety disorder.

AZD-6280 is a selective GABAA(α2/3) receptor modulator, used for treatment of generalized anxiety disorder.

Specifications

Materiale o nome chimico AZD-6280
CAS 942436-93-3
Colore Light Yellow
Note sul grado di purezza Research
Formula molecolare C20H22N4O3
Quantità 5 mg
Informazioni di solubilità DMSO : 62.5 mg/mL (170.57 mM; Need ultrasonic)
SMILES O=C(C1=NN=C2C(C3=CC(OC)=CC=C3OC)=CC=CC2=C1N)NCCC
Molecular Weight (g/mol) 366.41
Peso formulazione 366.41
Percent Purity 98.17%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.