Learn More
Azlon™ Multi-lingual Round Integral Wash Bottle
Descrizione
- Multilingual
- Wide neck wash bottle
- Indelibly printed in English, French, German and Spanish
- Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
- Fitted with color coded one-piece PP closure that maintains it's shape in use
- Designed with integral spout to prevent contamination
Specifica
Specifica
| Capacità (sistema metrico) | 500 mL |
| Include | Integral spout |
| Colore chiusura | Rosso |
| Closure Included | Sì |
| Bocca | Larga |
| Chemical Label | Acetone |
| Materiale chiusura | Polypropylene |
| Product Type | Wash Bottle |