missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam, 95.7%, MP Biomedicals™

Catalog Number p-4404661
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
78110-38-0
C13H17N5O8S2
435.426
WZPBZJONDBGPKJ-PSGLRMFWSA-N
Azthreonam, Azactam
56842327
2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
78110-38-0
C13H17N5O8S2
435.426
WZPBZJONDBGPKJ-PSGLRMFWSA-N
Azthreonam, Azactam
56842327
2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifically targets penicillin-binding protein 3 (PBP-3) of Gram-negative bacteria

  • Displays high degree of resistance to β-lactamase degradation
  • With high specificity for gram-negative aerobic rods
  • White Powder
Materiale o nome chimico Aztreonam
Tipo Aztreonam
Forma fisica Crystalline Powder
Intervallo percentuale saggio 0.957
Peso formulazione 435.4
Colore White
Quantità 1 g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'