Learn More
Brady™ BBP12 Label Printer Kit EU with Unwinder, Barcode Reader, and Workstation Scan and Print Suite
Product Code. 17135421 Shop All Brady Products
Change view
Click to view available options
BBP12 Label Printer EU Kit; Depth (Metric): 259 mm; Height (Metric): 191 mm; Width (Metric): 202 mm; Color Capability: Monochrome; Diameter (Metric): 25.4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17135421 BBP12 Label Printer EU Kit; Depth (Metric): 259 mm; Height (Metric): 191 mm; Width (Metric): 202 mm; Color Capability: Monochrome; Diameter (Metric): 25.4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17135421 Supplier Brady™ Supplier No. 302670
  • ONLINE EXCLUSIVE PRICE
    1580.00€
    1266.45€ / pezzi 1
    Risparmia 313.55€
    19% Sconto
Spedizione stimata: 09-12-2026
This practical kit includes printer, unwinder, barcode reader and Workstation Scan and Print Suite.
The BBP™12 Label Printer packs a lot of punch into one compact tool with its high-speed printing, extended list of labeling options and simple, user-friendly design.
  • Gives versatility to print all basic label types from one machine, saving costs and reducing downtime
  • Over 800 varieties of labels, including external-feed bulk rolls and custom parts for general identification, wire marking, asset tracking and more
  • Specifically designed small core media rolls and ribbons provide fast installation and reduce change-over time
  • Centering ribbon spindle virtually eliminates ribbon wrinkle
  • Compact size won't clutter your workplace or bench
  • Ships with unwinder, barcode reader, and the Brady™ Workstation Scan and Print application for label template design and manually triggered automated label printing
Specifications
Display Type Color LCD
Print Technology Thermal Transfer
Width (English) 8 in.
Width (Metric) 203 mm
Height (English) 7.5 in.
Height (Metric) 191 mm
Depth (English) 10 in.
Depth (Metric) 259 mm
Diameter (English) 1 in.
Diameter (Metric) 25.4 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes Printer, Unwinder, USB Cable, EU Power Cord, Product CD, Quick Start Guide, Xenon 1900HD Barcode Reader, Brady™ Workstation Scan and Print Suite
Languages Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian, Polish, Portuguese, Russian, Slovakian, Spanish, Swedish, Turkish
Memory 64 MB
Model BBP12
Print Speed 101.6 mm/s
Resolution 300 dpi
Operating System Windows™ 7, Windows™ 8, Windows™ 10
Temperature Range (Metric) 5°C to 40°C
Weight (English) 5.1 lb.
Weight (Metric) 2.3 kg
For Use With (Application) Barcode Labeling, Laboratory Labeling, Wire and Cable Labeling
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number BBP12-SPS-EU