Learn More
Berkshire International BCR™ Aluminium Flat Mop Handle
Codice prodotto. 18899954 Sfoglia Tutto Berkshire Prodotti
Cambia vista
Click to view available options
Materiale:
Anodized Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Materiale
18899954 Anodized Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18899954 Fornitore Berkshire International N. del fornitore BCR.T2HANDLE.10
  • ONLINE EXCLUSIVE PRICE
    840.00€
    646.28€ / pezzi 10
    Risparmia 193.72€
    23% Sconto
Spedizione stimata: 21-01-2027
Extremely manoeuvreable with a 360-degree swiveling head and telescopic handle extends from 95-180 cm.
  • Telescopic handle 95-180 cm
  • Anodized aluminum; lightweight and strong
  • Autoclavable (Recommended maximum of 121°C for 30 min)
  • Telescopic handle extends from 95-180 cm
Specifica
Product Type Cleanroom Mop Handle
Sterilità Non-sterile
Autoclavabile Autoclavable
Color-coded No
Materiale Anodized Aluminum