Learn More
BD Anti-rat Ig HRP Detection kit
Product Code. 15816478 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15816478 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15816478 Supplier BD Supplier No. 551013
  • ONLINE EXCLUSIVE PRICE
    795.00€
    675.79€ / 1 sets
    Risparmia 119.21€
    14% Sconto
 Restricted Item
Spedizione stimata: 26-11-2026
Includes
Biotinylated goat anti-rat Ig, Antibody Diluent Buffer, Streptavidin-Horseradish peroxidase (SAv-HRP), DAB Buffer, and DAB Chromogen
Designed for immunohistochemical staining of acetone-fixed frozen sections, formalin- or zinc-fixed paraffin sections, and cytospins/smears
  • Used with any purified primary antibody
  • Provides maximum reactivity
  • Contains protein stabilizers to enhance antibody reactivity
  • Blocking agents reduce non-specific staining
Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Storage Requirements 4°C
For Use With (Application) Immunohistochemistry, Immunohistochemical Staining
Quantity 1 Kit