Learn More
BD BBL™ Rabbit Coagulase Plasma
Product Code. 11965105
Change view
Click to view available options
Quantity:
10 x 3 mL
10 x 15 mL
Unit Size:
150mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11965105 10 x 3 mL Pack of 10
11758852 10 x 15 mL 150mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11965105 Supplier BD Supplier No. 240827
  • ONLINE EXCLUSIVE PRICE
    213.99€
    199.15€ / pezzi 10
    Risparmia 14.84€
    6% Sconto
 Restricted Item
Spedizione stimata: 09-12-2026
Qualitative determination of Staphylococci pathogenicity using direct tube method
Coagulase binds plasma fibrinogen, causing the organisms to agglutinate or plasma to clot. BBL Coagulase Plasma, Rabbit and BBL Coagulase Plasma Rabbit with EDTA are recommended for performing the direct tube test. At the time of use, test both positive and negative control cultures to check performance of the coagulase plasma.
  • Product type: diagnostic reagents and adjuvants
  • Groupings: C Products
  • Type: Coagulase Testing
  • Shelf Life: 1095 days
  • Label has Natural Rubber Latex statement
Certifications
CE marked
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE
Content And Storage 2°C to 8°C
Form Lyophilized
Product Type Coagulase Rabbit Plasma
For Use With (Application) Qualitative determination of Staphylococci pathogenicity using direct tube method
Quantity 10 x 3 mL
Species Rabbit