Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Streptomycin
Descrizione
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifica
Specifica
| Agenti antimicrobici | Streptomycin |
| Concentrazione | 300 μg |
| Tipo di imballaggio | Disc |
| Quantità | 1 Pack |
For in vitro diagnostic use